Fmoc-L-Gla(OtBu)2-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-γ-carboxy-Glu(OtBu)₂-OH, Fmoc-Gla(OtBu)₂-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-4-(tert-butoxycarbonyl)-5-oxopentanoic acid

4004771.0250 4004771.1000
B-1265.0250BAEA 1807.14 CAD
B-1265.0250BA B-1265.1000BA
Fmoc-L-Gla(OtBu)2-OH
Fmoc-L-Gla(OtBu)2-OH

Sequence: Fmoc-γ-carboxy-Glu(OtBu)₂-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.


Synonym(s): Fmoc-Gla(OtBu)₂-OH

Formula: C₂₉H₃₅NO₈
MW: 525.598 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00038770
CAS Number: 111662-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR