(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid
Supplier: BACHEM AMERICAS INC
Synonyms:
O-tert-Butyl-Nalpha-Fmoc-D-tyrosine, Nalpha-Fmoc-O-tert-butyl-D-tyrosine, Fmoc-D-Tyr(tBu)-OH, N-Fmoc-O-tert-butyl-D-tyrosine
4006924.0001
4006924.0005
4006924.0025
B-1260.0001BAEA
206.51
CAD
B-1260.0001BA
B-1260.0005BA
B-1260.0025BA
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(tert-butoxy)phenyl)propanoic acid
Sequence: Fmoc-D-Tyr(tBu)-OH
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Formula:
C₂₈H₂₉NO₅ MW: 459.5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065684 CAS Number: 118488-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...