FMOC-O-tert-butyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

4093144.0005 4093144.0025 4093144.0100
B-1255.0005BAEA 184.64 CAD
B-1255.0005BA B-1255.0025BA B-1255.0100BA
FMOC-O-tert-butyl-L-tyrosine
FMOC-O-tert-butyl-L-tyrosine

Sequence: Fmoc-Tyr(tBu)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₈H₂₉NO₅
Storage Temperature: Ambient
MDL Number: MFCD00037129
CAS Number: 71989-38-3
EINECS: 276-262-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR