Fmoc-D-Thr(tBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-D-threonine, Fmoc-D-Thr(tBu)-OH, (2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(tert-butoxy)butanoic acid, N-Fmoc-O-tert-butyl-D-threonine

4004993.0001 4004993.0005 4004993.0025
B-1250.0001BAEA 180.07 CAD
B-1250.0001BA B-1250.0005BA B-1250.0025BA
Fmoc-D-Thr(tBu)-OH
Fmoc-D-Thr(tBu)-OH

Sequence: Fmoc-D-Thr(tBu)-OH


Bachem also offers phosphothreonine derivatives.

Formula: C₂₃H₂₇NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00080292
CAS Number: 138797-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR