Fmoc-O-tert-butyl-D-serine

Supplier: BACHEM AMERICAS INC

4009509.0001 4009509.0005 4009509.0025
B-1240.0001BAEA 180.07 CAD
B-1240.0001BA B-1240.0005BA B-1240.0025BA
Fmoc-O-tert-butyl-D-serine
Fmoc-O-tert-butyl-D-serine

Sequence: Fmoc-D-Ser(tBu)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₂₂H₂₅NO₅
Storage Temperature: Ambient
MDL Number: MFCD00077071
CAS Number: 128107-47-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR