O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-O-tert-butyl-L-serine, N-a-Fmoc-O-t-butyl-L-serine, Fmoc-Ser(tBu)-OH, O-tert-Butyl-N-Fmoc-L-serine

4003086.0005 4003086.0025 4003086.0100
B-1235.0005BAEA 180.07 CAD
B-1235.0005BA B-1235.0025BA B-1235.0100BA
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine

Sequence: Fmoc-Ser(tBu)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.


To minimize the concomitant racemization, couplings of Fmoc-Ser(OtBu)-OH with uronium/aminium reagents should be performed in the presence of collidine in place of DIEA.

Formula: C₂₂H₂₅NO₅
MW: 383.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037127
CAS Number: 71989-33-8
EINECS: 276-260-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR