Fmoc-D-Ser(BSi)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((tert-butyldimethylsilyl)oxy)propanoic acid, Fmoc-D-Ser(TBDMS)-OH

4006507.0001 4006507.0005
B-1230.0001BAEA 608.66 CAD
B-1230.0001BA B-1230.0005BA
Fmoc-D-Ser(BSi)-OH
Fmoc-D-Ser(BSi)-OH

Sequence: Fmoc-D-Ser(BSi)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₂₄H₃₁NO₅Si
MW: 441.60 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08276730
CAS Number: 201210-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR