Fmoc-Ser(BSi)-OH

Supplier: BACHEM AMERICAS INC

4006501.0001 4006501.0005
B-1225.0001BAEA 490.38 CAD
B-1225.0001BA B-1225.0005BA
Fmoc-Ser(BSi)-OH
Fmoc-Ser(BSi)-OH

Sequence: Fmoc-Ser(BSi)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.


The BSi (or TBDMS) ether is somewhat more labile towards TFA than the tBu ether protection of the Ser hydroxyl moiety. Ser(TBDMS) can be cleaaved selectiviely in the presence of Boc/tBu with Bu₄N fluoride in DMF, but Fmoc is removed under these conditions as well.

Formula: C₂₄H₃₁NO₅Si
Storage Temperature: Freezer
MDL Number: MFCD02092931
CAS Number: 146346-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR