Fmoc-Cys(tBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-S-(tert-butyl)-L-cysteine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine, N-Fmoc-S-tert-butyl-L-cysteine, Fmoc-Cys(tBu)-OH

4003574.0005 4003574.0025 4003574.0100
B-1220.0005BAEA 180.83 CAD
B-1220.0005BA B-1220.0025BA B-1220.0100BA
Fmoc-Cys(tBu)-OH
Fmoc-Cys(tBu)-OH

Sequence: Fmoc-Cys(tBu)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).

Formula: C₂₂H₂₅NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00037130
CAS Number: 67436-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR