Fmoc-Tyr(Bzl)-OH

Supplier: BACHEM AMERICAS INC

4006928.0001 4006928.0005 4006928.0025
B-1215.0001BAEA 185.46 CAD
B-1215.0001BA B-1215.0005BA B-1215.0025BA
Fmoc-Tyr(Bzl)-OH
Fmoc-Tyr(Bzl)-OH

Sequence: Fmoc-Tyr(Bzl)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₃₁H₂₇NO₅
Storage Temperature: Ambient
MDL Number: MFCD00065682
CAS Number: 71989-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR