Fmoc-Ser(Bzl)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-O-benzyl-L-serine

4004135.0005 4004135.0025
B-1200.0005BAEA 238.17 CAD
B-1200.0005BA B-1200.0025BA
Fmoc-Ser(Bzl)-OH
Fmoc-Ser(Bzl)-OH

Sequence: Fmoc-Ser(Bzl)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₂₅H₂₃NO₅
MW: 417.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065674
CAS Number: 83792-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR