Fmoc-Tyr(tBu)-OPfp

Supplier: BACHEM AMERICAS INC

4011861.0001 4011861.0005
B-1195.0001BAEA 175.66 CAD
B-1195.0001BA B-1195.0005BA
Fmoc-Tyr(tBu)-OPfp
Fmoc-Tyr(tBu)-OPfp

Sequence: Fmoc-Tyr(tBu)-OPfp


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₃₄H₂₈F₅NO₅
Storage Temperature: Freezer
MDL Number: MFCD00077077
CAS Number: 86060-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR