Fmoc-Thr(tBu)-OPfp

Supplier: BACHEM AMERICAS INC

Synonyms: (2S,3R)-Perfluorophenyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tert-butoxy)butanoate

4011857.0001 4011857.0005
B-1185.0001BAEA 175.66 CAD
B-1185.0001BA B-1185.0005BA
Fmoc-Thr(tBu)-OPfp
Fmoc-Thr(tBu)-OPfp

Sequence: Fmoc-Thr(tBu)-OPfp


Bachem also offers phosphothreonine derivatives.

Formula: C₂₉H₂₆F₅NO₅
MW: 563.521 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00077076
CAS Number: 117088-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR