Fmoc-Pro-OPfp

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-L-proline pentafluorophenyl ester

4011852.0005 4011852.0025
B-1175.0005BAEA 283.5 CAD
B-1175.0005BA B-1175.0025BA
Fmoc-Pro-OPfp
Fmoc-Pro-OPfp

Sequence: Fmoc-Pro-OPfp

Formula: C₂₆H₁₈F₅NO₄
Storage Temperature: Freezer
MDL Number: MFCD00065672
CAS Number: 86060-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR