Fmoc-Lys(boc)-OPfp

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Lys(Boc)-OPfp

4011853.0001 4011853.0005
B-1155.0001BAEA 180.07 CAD
B-1155.0001BA B-1155.0005BA
Fmoc-Lys(boc)-OPfp
Fmoc-Lys(boc)-OPfp

Sequence: Fmoc-Lys(Boc)-OPfp


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₃₂H₃₁F₅N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00065661
CAS Number: 86060-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR