Fmoc-Gly-OPfp

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Gly-Opfp

4011859.0001 4011859.0005
B-1135.0001BAEA 175.66 CAD
B-1135.0001BA B-1135.0005BA
Fmoc-Gly-OPfp
Fmoc-Gly-OPfp

Sequence: Fmoc-Gly-OPfp


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₂₃H₁₄F₅NO₄
MW: 463.36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00037641
CAS Number: 86060-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR