5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Glu(OtBu)-OPfp, Fmoc-Glu(OtBu)-Opfp, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, N-Fmoc-L-glutamic Acid 5-tert-Butyl 1-Pentafluorophenyl Ester, 5-tert-Butyl 1-Pentafluorophenyl N-Fmoc-L-glutamate, N-Fmoc-L-glutamic acid 5-tert-butyl ester pentafluorophenyl ester

4011874.0005 4011874.0025
B-1130.0005BAEA 601.51 CAD
B-1130.0005BA B-1130.0025BA
5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate
5-tert-Butyl-1-pentafluorophenyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-glutamate

Sequence: Fmoc-Glu(OtBu)-OPfp


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₃₀H₂₆F₅NO₆
MW: 591.53 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00065647
CAS Number: 86061-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR