(R)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tritylthio)propanoate
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-Cys(Trt)-OPfp, N-Fmoc-S-trityl-L-cysteine pentafluorophenyl ester
4030624.0001
4030624.0005
4030624.0025
B-1120.0001BAEA
175.66
CAD
B-1120.0001BA
B-1120.0005BA
B-1120.0025BA
(R)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tritylthio)propanoate
(R)-Perfluorophenyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(tritylthio)propanoate
Sequence: Fmoc-Cys(Trt)-OPfp
Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).
Formula:
C₄₃H₃₀F₅NO₄S MW: 751.768 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00065639 CAS Number: 115520-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...