Fmoc-Asn-OPfp

Supplier: BACHEM AMERICAS INC

4011838.0005 4011838.0025
B-1105.0005BAEA 400.8 CAD
B-1105.0005BA B-1105.0025BA
Fmoc-Asn-OPfp
Fmoc-Asn-OPfp

Sequence: Fmoc-Asn-OPfp


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₂₅H₁₇F₅N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00065625
CAS Number: 86060-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR