Fmoc-D-Orn(Boc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nδ-Boc-Nα-Fmoc-D-ornithine

4004451.0001 4004451.0005 4004451.0025
B-1095.0001BAEA 206.51 CAD
B-1095.0001BA B-1095.0005BA B-1095.0025BA
Fmoc-D-Orn(Boc)-OH
Fmoc-D-Orn(Boc)-OH

Sequence: Fmoc-D-Orn(Boc)-OH


Bachem also offers Fmoc-Orn(Boc)-Wang resin (D-1800).

Formula: C₂₅H₃₀N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00077065
CAS Number: 118476-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR