Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine

Supplier: BACHEM AMERICAS INC

Synonyms: Nδ-Boc-Nɑ-Fmoc-L-ornithine, Nδ-Boc-Nα-Fmoc-L-ornithine, Fmoc-Orn(Boc)-OH

4004449.0005 4004449.0025 4004449.0100
B-1090.0005BAEA 206.51 CAD
B-1090.0005BA B-1090.0025BA B-1090.0100BA
Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine
Ndelta-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine

Sequence: Fmoc-Orn(Boc)-OH


Bachem also offers Fmoc-Orn(Boc)-Wang resin (D-1800).

Formula: C₂₅H₃₀N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00065668
CAS Number: 109425-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR