Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine

Supplier: BACHEM AMERICAS INC

Synonyms: Nε-Boc-Nɑ-Fmoc-D-lysine, Fmoc-D-Lys(Boc)-OH

4004439.0005 4004439.0025 4004439.0100
B-1085.0005BAEA 175.66 CAD
B-1085.0005BA B-1085.0025BA B-1085.0100BA
Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine
Nepsilon-(tert-butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine

Sequence: Fmoc-D-Lys(Boc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065660
CAS Number: 92122-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR