N-ɑ-Fmoc-N-ε-boc-L-lysine

Supplier: BACHEM AMERICAS INC

Synonyms: N-ε-(tert-Butoxycarbonyl)-N-ɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid, N-ε-Boc-N-ɑ-Fmoc-L-lysine, Fmoc-Lys(Boc)-OH

4003151.0025 4003151.0100 4003151.0500
B-1080.0025BAEA 290.81 CAD
B-1080.0025BA B-1080.0100BA B-1080.0500BA
N-ɑ-Fmoc-N-ε-boc-L-lysine
N-alpha-Fmoc-N-epsilon-boc-L-lysine

Sequence: Fmoc-Lys(Boc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037138
CAS Number: 71989-26-9
EINECS: 276-256-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR