(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Supplier: BACHEM AMERICAS INC

4006402.0001 4006402.0005 4006402.0025
B-1070.0001BAEA 201.47 CAD
B-1070.0001BA B-1070.0005BA B-1070.0025BA
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

Sequence: Fmoc-D-Asp(OtBu)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₃H₂₅NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00077050
CAS Number: 112883-39-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR