4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate

Supplier: BACHEM AMERICAS INC

4006808.0005 4006808.0025
B-1060.0005BAEA 291.08 CAD
B-1060.0005BA B-1060.0025BA
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate

Sequence: Fmoc-Asp(OBzl)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₆H₂₃NO₆
Storage Temperature: Ambient
MDL Number: MFCD00065630
CAS Number: 86060-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR