Fmoc-Arg-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Fmoc-L-arginine

4003163.0005 4003163.0025 4003163.0100
B-1040.0005BAEA 185.46 CAD
B-1040.0005BA B-1040.0025BA B-1040.0100BA
Fmoc-Arg-OH
Fmoc-Arg-OH

Sequence: Fmoc-Arg-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₁H₂₄N₄O₄
MW: 396.447 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00051770
CAS Number: 91000-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR