N-Fmoc-β-alanine
Supplier: BACHEM AMERICAS INC
Synonyms:
Fmoc-β-Ala-OH P, Fmoc-β-Ala-p-alkoxybenzyl alcohol, N-Fmoc-beta-alanine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-beta-alanine
4007989.0005
4007989.0025
4007989.0100
B-1025.0005BAEA
185.46
CAD
B-1025.0005BA
B-1025.0025BA
B-1025.0100BA
N-Fmoc-β-alanine
N-Fmoc-β-alanine
Sequence: Fmoc-β-Ala-OH
Bachem also offers Fmoc-β-Ala-Wang resin (D-1625) and Boc-β-Ala-PAM resin (D-1650).
Formula:
C₁₈H₁₇NO₄ MW: 311.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00063328 CAS Number: 35737-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...