Fmoc-Ser(Ac)-OH

Supplier: BACHEM AMERICAS INC

4009560.0001 4009560.0005
B-1010.0001BAEA 232.35 CAD
B-1010.0001BA B-1010.0005BA
Fmoc-Ser(Ac)-OH
Fmoc-Ser(Ac)-OH

Sequence: Fmoc-Ser(Ac)-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₂₀H₁₉NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00077070
CAS Number: 171778-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR