Fmoc-Cys(Acm)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Fmoc-S-acetamidomethyl-L-cysteine

4006622.0005 4006622.0025 4006622.0100
B-1005.0005BAEA 180.14 CAD
B-1005.0005BA B-1005.0025BA B-1005.0100BA
Fmoc-Cys(Acm)-OH
Fmoc-Cys(Acm)-OH

Sequence: Fmoc-Cys(Acm)-OH


Besides "standard" preloaded resins, Bachem offers laterally anchored Fmoc-cysteine, Fmoc-Cys(SASRIN resin)-OH (D-2170) and resin-linked Fmoc-cysteamine, Fmoc-cysteamine-SASRIN resin (D-2165).


Following Fmoc-SPPS, Acm can be converted on resin into the S-carbomethoxysulfenyl (Scm) group. Scm is cleaved by reduction with DTT, treatment with thiols yields unsymmetrical disulfides.

Formula: C₂₁H₂₂N₂O₅S
Storage Temperature: Ambient
MDL Number: MFCD00038769
CAS Number: 86060-81-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR