(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

Supplier: BACHEM AMERICAS INC

4039941.0250 4039941.1000
A-4750.0250BAEA 309.81 CAD
A-4750.0250BA A-4750.1000BA
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid
(2S,4S)-1-(tert-Butoxycarbonyl)-4-fluoro-2-pyrrolidinecarboxylic acid

Sequence: Boc-cis-4-fluoro-Pro-OH


Flp derivatives.


Synonym(s): Boc-(2S,4S)-4-fluoro-pyrrolidine-2-carboxylic acid#Boc-flp-OH

Formula: C₁₀H₁₆FNO₄
Storage Temperature: Freezer
MDL Number: MFCD04973957
CAS Number: 203866-13-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR