N-(tert-Butoxycarbonyl)-L-propargylglycine

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-ynoic acid, N-Boc-L-propargylglycine, (S)-N-BOC-Propargylglycine, Boc-propargyl-Gly-OH, (S)-2-(Boc-amino)-4-pentynoic Acid, Boc-Pra-OH, (S)-2-(tert-Butoxycarbonylamino)-4-pentynoic Acid

4038616.0001 4038616.0005
A-4735.0001BAEA 678.52 CAD
A-4735.0001BA A-4735.0005BA
N-(tert-Butoxycarbonyl)-L-propargylglycine
N-(tert-Butoxycarbonyl)-L-propargylglycine

Sequence: Boc-Pra-OH
Synonym(s): Boc-Propargyl-Gly-OH#(S)-2-(Boc-amino)-4-pentynoic acid


Bachem also offers resin-bound propargylglycine, Fmoc-Pra-Wang resin (D-2820).


Used in the synthesis of neoglycopeptides employing chemical ligation and click chemistry.

Formula: C₁₀H₁₅NO₄
MW: 213.23 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01320855
CAS Number: 63039-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR