ZINC34456714

Supplier: BACHEM AMERICAS INC

4050107.0001 4050107.0005
A-4715.0001BAEA 624.24 CAD
A-4715.0001BA A-4715.0005BA
ZINC34456714
ZINC34456714

Sequence: Boc-Dap-OtBu


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.

Formula: C₁₂H₂₄N₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD08275854
CAS Number: 77215-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR