[(tert-Butoxycarbonyl)aminooxy]acetic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-(carboxymethoxy)amine, Boc-Aoa-OH, (Boc-aminooxy)acetic Acid

4038799.0001 4038799.0005
A-4605.0001BAEA 180.91 CAD
A-4605.0001BA A-4605.0005BA
[(tert-Butoxycarbonyl)aminooxy]acetic acid
[(tert-Butoxycarbonyl)aminooxy]acetic acid

Sequence: Boc-AOAc-OH
Synonym(s): 2-(Boc-aminooxy)-acetic acid#N-Boc-(carboxymethoxy)-amine


Protected aminooxyacetic acid derivative for the incorporation of an N-terminal O-alkylhydroxylamine moiety, which is used in the chemoselective ligation of unprotected peptide fragments, for the conjugation to lipids and glycosides, and for the introduction of labels by oxime formation. Peptides containing AOAc may be cleaved selectively at the N-O bond with Zn/AcOH. See also the 12-aminooxydodecanoic acid derivative A-4720.

Formula: C₇H₁₃NO₅
MW: 191.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01632027
CAS Number: 42989-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR