Boc-L-alaninal

Supplier: BACHEM AMERICAS INC

4033832.0001 4033832.0005
A-4590.0001BAEA 452.5 CAD
A-4590.0001BA A-4590.0005BA
Boc-L-alaninal
Boc-L-alaninal

Sequence: Boc-Ala-aldehyde


Boc-L-alaninal.

Formula: C₈H₁₅NO₃
Storage Temperature: Freezer
MDL Number: MFCD00143786
CAS Number: 79069-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR