Boc-D-Chg-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-D-Chg-OH, N-(tert-Butoxycarbonyl)-D-2-cyclohexylglycine, N-Boc-2-cyclohexyl-D-glycine, Boc-cyclohexyl-D-Gly-OH, (R)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid, BOC-D-α-cyclohexylglycine, N-Boc-D-2-cyclohexylglycine
Sequence: Boc-cyclohexyl-D-Gly-OH
Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.
Chg derivatives.
Synonym(s): Boc-D-Chg-OH
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00133629 CAS Number: 70491-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...