Boc-Gly-Gly-Phe-Gly-OH
Supplier: BACHEM AMERICAS INC
4028365.0250 4028365.1000
A-4450.0250BAEA 264.68 CAD
A-4450.0250BA A-4450.1000BA
Boc-Gly-Gly-Phe-Gly-OH
Proteins and Peptides
For Boc-dipeptides Boc-Xaa-Gly-OH and Boc-Xaa-Pro-OH, which can be used as building blocks during Boc-SPPS, please see the product family 'Boc-Dipeptide Building Blocks' in the 'Amino Acid Derivatives' section. This family also includes short protected peptide fragments suitable for solution synthesis and preparation of enzyme substrates and inhibitors.