Boc-N-Me-D-Tyr-OH·DCHA
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-N-Me-D-Tyr-OH·DCHA
4028209.0001
4028209.0005
A-4420.0001BAEA
593.81
CAD
A-4420.0001BA
A-4420.0005BA
Boc-N-Me-D-Tyr-OH·DCHA
Boc-N-Me-D-Tyr-OH·DCHA
Sequence: Boc-N-Me-D-Tyr-OH · DCHA
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Formula:
C₂₇H₄₄N₂O₅ Storage Temperature: Ambient |
MDL Number:
MFCD02259474 CAS Number: 178208-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...