Boc-Dab-OtBu·HCl
Supplier: BACHEM AMERICAS INC
4028125.0001
4028125.0005
A-4415.0001BAEA
593.81
CAD
A-4415.0001BA
A-4415.0005BA
Boc-Dab-OtBu·HCl
Boc-Dab-OtBu·HCl
Sequence: Boc-Dab-OtBu · HCl
Formula:
C₁₃H₂₆N₂O₄ MW: 274.361 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD02259473 CAS Number: 190447-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...