Boc-Hyp-OtBu

Supplier: BACHEM AMERICAS INC

Synonyms: (2S,4R)-Di-tert-Butyl 4-hydroxypyrrolidine-1,2-dicarboxylate

4027945.0001 4027945.0005
A-4405.0001BAEA 387.21 CAD
A-4405.0001BA A-4405.0005BA
Boc-Hyp-OtBu
Boc-Hyp-OtBu

Sequence: Boc-Hyp-OtBu


Hyp denotes L-trans-hydroxyproline.

Formula: C₁₄H₂₅NO₅
Storage Temperature: Freezer
MDL Number: MFCD01318729
CAS Number: 170850-75-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR