N-Boc-N'-Fmoc-diaminoacetic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-α-amino-DL-Gly(Boc)-OH

4027508.0001 4027508.0005
A-4320.0001BAEA 516.34 CAD
A-4320.0001BA A-4320.0005BA
N-Boc-N'-Fmoc-diaminoacetic acid
N-Boc-N'-Fmoc-diaminoacetic acid

Sequence: Fmoc-α-amino-DL-Gly(Boc)-OH


Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.


Synonym(s): Fmoc-DL-Agl(Boc)-OH#Boc-α-amino-DL-Gly(Fmoc)-OH#(RS)-2-(Boc-amino)-2-(Fmoc-amino)-acetic acid

Formula: C₂₂H₂₄N₂O₆
MW: 412.442 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00273452
CAS Number: 176039-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR