Boc-D-Dap(Z)-OH·DCHA
Supplier: BACHEM AMERICAS INC
Synonyms:
Dicyclohexylamine (R)-3-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)propanoate
4027600.0001
4027600.0005
A-4265.0001BAEA
541.99
CAD
A-4265.0001BA
A-4265.0005BA
Boc-D-Dap(Z)-OH·DCHA
Boc-D-Dap(Z)-OH·DCHA
Sequence: Boc-D-Dap(Z)-OH · DCHA
Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.
Formula:
C₂₈H₄₅N₃O₆ MW: 519.683 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798629 CAS Number: 81306-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...