Boc-D-Lys(fmoc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Boc-Nepsilon-Fmoc-D-lysine, Boc-D-Lys(Fmoc)-OH

4030412.0001 4030412.0005
A-3985.0001BAEA 529.57 CAD
A-3985.0001BA A-3985.0005BA
Boc-D-Lys(fmoc)-OH
Boc-D-Lys(fmoc)-OH

Sequence: Boc-D-Lys(Fmoc)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076966
CAS Number: 115186-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR