Boc-4-(Fmoc-amino)-1-phenylalanine

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-3-(4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid, Boc-p-amino-Phe(Fmoc)-OH, Boc-Phe(4-NHFmoc)-OH, N-Boc-4-(Fmoc-amino)-L-phenylalanine

4026806.0001 4026806.0005
A-3975.0001BAEA 206.51 CAD
A-3975.0001BA A-3975.0005BA
Boc-4-(Fmoc-amino)-1-phenylalanine
Boc-4-(Fmoc-amino)-1-phenylalanine

Sequence: Boc-p-amino-Phe(Fmoc)-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Aph derivatives.

Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151889
CAS Number: 114346-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR