Boc-Thz-OH

Supplier: BACHEM AMERICAS INC

4029316.0005 4029316.0025
A-3945.0005BAEA 183.09 CAD
A-3945.0005BA A-3945.0025BA
Boc-Thz-OH
Boc-Thz-OH

Sequence: Boc-L-thiazolidine-4-carboxylic acid


Thiazolidine-4-carboxylic acid (Thz) can be inserted as a Pro equivalent (thiaproline) as well as a masked cysteine. Contrary to Cys derivatives, Thz derivatives can be activated and coupled with minimal racemization.


Synonym(s): Boc-Thz-OH#Boc-L-thioproline

Formula: C₉H₁₅NO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00077002
CAS Number: 51077-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR