N-(tert-Butoxycarbonyl)-L-pyroglutamic acid

Supplier: BACHEM AMERICAS INC

4026369.0001 4026369.0005 4026369.0025
A-3850.0001BAEA 180.91 CAD
A-3850.0001BA A-3850.0005BA A-3850.0025BA
N-(tert-Butoxycarbonyl)-L-pyroglutamic acid
N-(tert-Butoxycarbonyl)-L-pyroglutamic acid

Sequence: Boc-Pyr-OH


Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.

Formula: C₁₀H₁₅NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00672316
CAS Number: 53100-44-0
EINECS: 258-362-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR