Boc-L-HArg(Et)2-OH

Supplier: BACHEM AMERICAS INC

4026054.0050 4026054.0250
A-3775.0050BAEA 593.81 CAD
A-3775.0050BA A-3775.0250BA
Boc-L-HArg(Et)2-OH
Boc-L-HArg(Et)2-OH

Sequence: Boc-Homoarg(Et)₂-OH (symmetrical)


Har derivatives.


Nω,ω'-alkylated derivative of homoarginine. Building block for LHRH antagonists as Ganirelix, which contains both enantiomers of Har(Et)₂.

Formula: C₁₆H₃₂N₄O₄
Storage Temperature: Freezer
MDL Number: MFCD18782098
CAS Number: 122532-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR