Boc-Cha-OH

Supplier: BACHEM AMERICAS INC

4025893.0001 4025893.0005
A-3760.0001BAEA 206.51 CAD
A-3760.0001BA A-3760.0005BA
Boc-Cha-OH
Boc-Cha-OH

Sequence: Boc-β-cyclohexyl-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Cha corresponds to hydrogenated phenylalanine, the aromatic nucleus is replaced by a more flexible aliphatic cycle.


Synonym(s): Boc-Cha-OH

Formula: C₁₄H₂₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00076896
CAS Number: 37736-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR