Boc-Orn-OtBu·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Orn-OtBu·HCl, (S)-tert-Butyl 5-amino-2-((tert-butoxycarbonyl)amino)pentanoate hydrochloride

4025480.0001 4025480.0005
A-3755.0001BAEA 352.81 CAD
A-3755.0001BA A-3755.0005BA
Boc-Orn-OtBu·HCl
Boc-Orn-OtBu·HCl

Sequence: Boc-Orn-OtBu · HCl

Formula: C₁₄H₂₉ClN₂O₄
MW: 324.846 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01318721
CAS Number: 214629-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR