Boc-D-allo-Isoleucine
Supplier: BACHEM AMERICAS INC
Synonyms:
Boc-D-allo-Ile-OH, (2R,3S)-2-((tert-Butoxycarbonyl)amino)-3-methylpentanoic acid
4025635.0001
4025635.0005
A-3735.0001BAEA
672.04
CAD
A-3735.0001BA
A-3735.0005BA
Boc-D-allo-Isoleucine
Boc-D-allo-Isoleucine
Sequence: Boc-D-allo-Ile-OH
Formula:
C₁₁H₂₁NO₄ MW: 231.292 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00237543 CAS Number: 55780-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...