Boc-N-Me-Lys(Z)-OH · DCHA

Supplier: BACHEM AMERICAS INC

4025309.0001 4025309.0005
A-3690.0001BAEA 361.41 CAD
A-3690.0001BA A-3690.0005BA
Boc-N-Me-Lys(Z)-OH · DCHA
Boc-N-Me-Lys(Z)-OH · DCHA

Sequence: Boc-N-Me-Lys(Z)-OH · DCHA


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₀H₃₀N₂O₆·C₁₂H₂₃N
Storage Temperature: Refrigerator
MDL Number: MFCD00236866
CAS Number: 201002-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR